Anti-CRYM Mouse monoclonal antibody [clone: 10B2609]

USBIC7944-93EA 0 EUR
USBIC7944-93
Anti-CRYM Mouse monoclonal antibody [clone: 10B2609]
Type: Primary
Antigen: CRYM (crystallin, mu)
Clonality: Monoclonal
Clone: 10B2609
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR