Anti-CRYAB Goat polyclonal antibody

Lieferant: United States Biological
USBIC7944-60CEA 640 EUR
USBIC7944-60C
Anti-CRYAB Goat polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR