Anti-Cryptosporidium sp. Mouse monoclonal antibody [clone: 10B2304]

Lieferant: United States Biological
USBIC7944-22EA 538 EUR
USBIC7944-22
Anti-Cryptosporidium sp. Mouse monoclonal antibody [clone: 10B2304]
Antikörper
Recommended dilution: ELISA: 1:2000-1:8000; Immunofluorescence: 1:50-1:200

Type: Primary
Antigen: Cryptosporidium sp.
Clonality: Monoclonal
Clone: 10B2304
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR