Anti-Cryptosporidium parvum Goat polyclonal antibody

Lieferant: United States Biological
USBIC7944-16EA 374 EUR
USBIC7944-16
Anti-Cryptosporidium parvum Goat polyclonal antibody
Antikörper
Recommended dilution: Immunofluorescence: 1:1000

Type: Primary
Antigen: Cryptosporidium parvum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR