Anti-Calcitonin Receptor-stimulating Peptide 3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC7936-65BEA 1090 EUR
USBIC7936-65B
Anti-Calcitonin Receptor-stimulating Peptide 3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Calcitonin Receptor-stimulating Peptide 3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR