Anti-CREB5 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2600]

USBIC7915-52-APEA 0 EUR
USBIC7915-52-AP
Anti-CREB5 Mouse monoclonal antibody AP (Alkaline Phosphatase) [clone: 10B2600]
Type: Primary
Antigen: CREB5 (cAMP responsive element binding protein 5)
Clonality: Monoclonal
Clone: 10B2600
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR