Anti-CREB Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC7915-05KEA 347 EUR
USBIC7915-05K
Anti-CREB Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot:1:500-1:1000; Immunochemistry:1:50-1:100

Type: Primary
Antigen: CREB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR