Anti-CREB Rabbit monoclonal antibody [clone: 5i117]

Lieferant: United States Biological
USBIC7915-04EA 501 EUR
USBIC7915-04
Anti-CREB Rabbit monoclonal antibody [clone: 5i117]
Antikörper
Recommended dilution: Flow Cytometry: 1:50; Western Blot: 1:1000; Immunochemistry: 1:40

Type: Primary
Antigen: CREB
Clonality: Monoclonal
Clone: 5i117
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR