Anti-CKM Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC7910-24A-BIOTEA 679 EUR
USBIC7910-24A-BIOT
Anti-CKM Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CKM (creatine kinase, muscle)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR