Anti-CKMT2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC7910-21A-BIOTEA 679 EUR
USBIC7910-21A-BIOT
Anti-CKMT2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CKMT2 (creatine kinase, mitochondrial 2 (sarcomeric))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR