Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 10K174]

Lieferant: United States Biological
USBIC7907-10BEA 481 EUR
USBIC7907-10B
Anti-C-Reactive Protein Mouse monoclonal antibody [clone: 10K174]
Antikörper
Type: Primary
Antigen: C-Reactive Protein
Clonality: Monoclonal
Clone: 10K174
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR