Anti-CRAMP Rabbit polyclonal antibody Carboxyfluorescein

Lieferant: United States Biological
USBIC7906-74M2EA 741 EUR
USBIC7906-74M2
Anti-CRAMP Rabbit polyclonal antibody Carboxyfluorescein
Antikörper
Type: Primary
Antigen: CRAMP
Clonality: Polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR