Anti-CRAMP Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC7906-74MEA 585 EUR
USBIC7906-74M
Anti-CRAMP Rabbit polyclonal antibody
Antikörper
Type: Primary
Antigen: CRAMP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR