Anti-CAMP Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIC7906-74CEA 0 EUR
USBIC7906-74C
Anti-CAMP Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CAMP (cathelicidin antimicrobial peptide)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR