Anti-CRABP2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC7906-71GEA 507 EUR
USBIC7906-71G
Anti-CRABP2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-2?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CRABP2 (cellular retinoic acid binding protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR