Anti-CPT1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIC7906-06A1-APCEA 711 EUR
USBIC7906-06A1-APC
Anti-CPT1A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!

Type: Primary
Antigen: CPT1A (carnitine palmitoyltransferase 1A (liver))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR