Anti-COX6B1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2586]

USBIC7904-89L-APCEA 0 EUR
USBIC7904-89L-APC
Anti-COX6B1 Mouse monoclonal antibody APC (Allophycocyanin) [clone: 10B2586]
Type: Primary
Antigen: COX6B1 (cytochrome c oxidase subunit VIb polypeptide 1 (ubiquitous))
Clonality: Monoclonal
Clone: 10B2586
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR