Anti-COX4I1 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC7904-89B1EA 578 EUR
USBIC7904-89B1
Anti-COX4I1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000; Western Blot: 1:4000; Immunochemistry: 2.5 ?g/ml

Type: Primary
Antigen: COX4I1 (cytochrome c oxidase subunit IV isoform 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR