Anti-COX 2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC7904-80QEA 448 EUR
USBIC7904-80Q
Anti-COX 2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COX 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR