Anti-CRHR1 Goat polyclonal antibody

Lieferant: United States Biological
USBIC7903-79C1EA 471 EUR
USBIC7903-79C1
Anti-CRHR1 Goat polyclonal antibody
Antikörper
Recommended dilution: ELISA: 1:16000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: CRHR1 (corticotropin releasing hormone receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR