Anti-Cornichon Homolog 4 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 11C275]

USBIC7901-06H-PEEA 0 EUR
USBIC7901-06H-PE
Anti-Cornichon Homolog 4 Mouse monoclonal antibody PE (Phycoerythrin) [clone: 11C275]
Type: Primary
Antigen: Cornichon Homolog 4
Clonality: Monoclonal
Clone: 11C275
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR