Anti-COPS5 Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIC7900-58AEA 793 EUR
USBIC7900-58A
Anti-COPS5 Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: COPS5 (COP9 signalosome subunit 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR