Anti-GJA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: United States Biological
USBIC7856-02J-APEA 852 EUR
USBIC7856-02J-AP
Anti-GJA5 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: GJA5 (gap junction protein, alpha 5, 40kDa)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR