Anti-GJA4 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIC7855-35B-FITCEA 0 EUR
USBIC7855-35B-FITC
Anti-GJA4 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: FLISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: GJA4 (gap junction protein, alpha 4, 37kDa)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR