Anti-COMT Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC7851-70KEA 578 EUR
USBIC7851-70K
Anti-COMT Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:4000; Western Blot: 1:4000; Immunochemistry: 3.75 ?g/ml

Type: Primary
Antigen: COMT (catechol-O-methyltransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR