Anti-CPLX2 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC7851-03EA 722 EUR
USBIC7851-03
Anti-CPLX2 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml

Type: Primary
Antigen: CPLX2 (complexin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR