Anti-Complement Factor H-Related Protein 1 Sheep Polyclonal Antibody

Lieferant: United States Biological
USBIC7850-86EA 777 EUR
USBIC7850-86
Anti-Complement Factor H-Related Protein 1 Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 0.5-1.0 ?g/ml; Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Complement Factor H-Related Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR