Anti-CFB Mouse Monoclonal Antibody (Biotin) [clone: 6F81]

Lieferant: United States Biological
USBIC7850-63BEA 856 EUR
USBIC7850-63B
Anti-CFB Mouse Monoclonal Antibody (Biotin) [clone: 6F81]
Antikörper
Type: Primary
Antigen: CFB (complement factor B)
Clonality: Monoclonal
Clone: 6F81
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR