Anti-C6 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC7850-35BEA 697 EUR
USBIC7850-35B
Anti-C6 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Western Blot: 1:10; Immunochemistry: 1:10

Type: Primary
Antigen: C6 (complement component 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR