Anti-C5b-9 Terminal Complement Complex Mouse monoclonal antibody [clone: 13A96]

Lieferant: United States Biological
USBIC7850-27BEA 958 EUR
USBIC7850-27B
Anti-C5b-9 Terminal Complement Complex Mouse monoclonal antibody [clone: 13A96]
Antikörper
Type: Primary
Antigen: C5b-9 Terminal Complement Complex
Clonality: Monoclonal
Clone: 13A96
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR