Anti-C5 Mouse monoclonal antibody [clone: 6F72]

Lieferant: United States Biological
USBIC7850-23AEA 671 EUR
USBIC7850-23A
Anti-C5 Mouse monoclonal antibody [clone: 6F72]
Antikörper
Type: Primary
Antigen: C5 (complement component 5)
Clonality: Monoclonal
Clone: 6F72
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR