Anti-Complement C3, C3c Sheep Polyclonal Antibody (FITC (Fluorescein))

Lieferant: United States Biological
USBIC7850-14REA 838 EUR
USBIC7850-14R
Anti-Complement C3, C3c Sheep Polyclonal Antibody (FITC (Fluorescein))
Antikörper
Recommended dilution: Immunochemistry: 1:50-1:100

Type: Primary
Antigen: Complement C3, C3c
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR