Anti-COMP Mouse Monoclonal Antibody [clone: 9H100]
Lieferant: United States Biological
USBIC7849-09EA 742 EUR
USBIC7849-09
Anti-COMP Mouse Monoclonal Antibody [clone: 9H100]
Antikörper
Recommended dilution: Western Blot: 2?g/ml
Type: Primary
Antigen: COMP (cartilage oligomeric matrix protein)
Clonality: Monoclonal
Clone: 9H100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human