Anti-COMMD1 Mouse monoclonal antibody [clone: 10B2565]

USBIC7847-50EA 0 EUR
USBIC7847-50
Anti-COMMD1 Mouse monoclonal antibody [clone: 10B2565]
Type: Primary
Antigen: COMMD1 (copper metabolism (Murr1) domain containing 1)
Clonality: Monoclonal
Clone: 10B2565
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR