Anti-COL5 Goat Polyclonal Antibody

Lieferant: United States Biological
USBIC7510-56KEA 708 EUR
USBIC7510-56K
Anti-COL5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COL5 (collagen, type V)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR