Anti-COL4 Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIC7510-54BEA 434 EUR
USBIC7510-54B
Anti-COL4 Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: COL4 (collagen, type IV)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: alpha 3 Binding Protein
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR