Anti-COL3 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC7510-44LEA 448 EUR
USBIC7510-44L
Anti-COL3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunochemistry: 1:20

Type: Primary
Antigen: COL3 (collagen, type III)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR