Anti-CNPPD1 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBIC6206-01EA 519 EUR
USBIC6206-01
Anti-CNPPD1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:500-1:3000; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CNPPD1 (cyclin Pas1/PHO80 domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR