Anti-Clostridium difficile Toxin A Mouse Monoclonal Antibody [clone: 10B2261]

Lieferant: United States Biological
USBIC5851-95JEA 513 EUR
USBIC5851-95J
Anti-Clostridium difficile Toxin A Mouse Monoclonal Antibody [clone: 10B2261]
Antikörper
Recommended dilution: ELISA: 1:20-1:200

Type: Primary
Antigen: Clostridium difficile Toxin A
Clonality: Monoclonal
Clone: 10B2261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR