Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 8B9]

Lieferant: United States Biological
USBIC5851-82EEA 671 EUR
USBIC5851-82E
Anti-Clostridium botulinum E Toxoid Mouse Monoclonal Antibody [clone: 8B9]
Antikörper
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: Clostridium botulinum E Toxoid
Clonality: Monoclonal
Clone: 8B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR