Anti-Clostridium botulinum Neurotoxin Type D Light Chain Sheep Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC5851-49GEA 688 EUR
USBIC5851-49G
Anti-Clostridium botulinum Neurotoxin Type D Light Chain Sheep Polyclonal Antibody (Biotin)
Antikörper
Recommended dilution: Western Blot: 0.1-0.2?g/ml

Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type D Light Chain
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR