Anti-Clostridium botulinum Neurotoxin Type B Chicken Polyclonal Antibody

Lieferant: United States Biological
USBIC5851-48TEA 441 EUR
USBIC5851-48T
Anti-Clostridium botulinum Neurotoxin Type B Chicken Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:42000-1:66000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Clostridium botulinum Neurotoxin Type B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR