Anti-Clostridium botulinum A Toxin Mouse Monoclonal Antibody [clone: 8J155]

Lieferant: United States Biological
USBIC5851-40DEA 455 EUR
USBIC5851-40D
Anti-Clostridium botulinum A Toxin Mouse Monoclonal Antibody [clone: 8J155]
Antikörper
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: Clostridium botulinum A Toxin
Clonality: Monoclonal
Clone: 8J155
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR