Anti-CLN5 Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBIC5841-80-BIOTEA 679 EUR
USBIC5841-80-BIOT
Anti-CLN5 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: CLN5 (ceroid-lipofuscinosis, neuronal 5)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR