Anti-CLEC14A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K67]

Lieferant: United States Biological
USBIC5839-73T1EA 619 EUR
USBIC5839-73T1
Anti-CLEC14A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 12K67]
Antikörper
Do not freeze!

Type: Primary
Antigen: CLEC14A (C-type lectin domain family 14, member A)
Clonality: Monoclonal
Clone: 12K67
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR