Anti-CPSF6 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2591]

USBIC5839-33-HRPEA 0 EUR
USBIC5839-33-HRP
Anti-CPSF6 Mouse monoclonal antibody HRP (Horseradish Peroxidase) [clone: 10B2591]
Type: Primary
Antigen: CPSF6 (cleavage and polyadenylation specific factor 6, 68kDa)
Clonality: Monoclonal
Clone: 10B2591
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR