Anti-CLDN5 Mouse monoclonal antibody [clone: 6A44]

Lieferant: United States Biological
USBIC5838-47BEA 544 EUR
USBIC5838-47B
Anti-CLDN5 Mouse monoclonal antibody [clone: 6A44]
Antikörper
Recommended dilution: ELISA: 0.1-1 ?g/ml; Western Blot: 1-3?g/ml; Immunochemistry: 5-10 ?g/ml

Type: Primary
Antigen: CLDN5 (claudin 5)
Clonality: Monoclonal
Clone: 6A44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR