Anti-CLDN3 Rabbit polyclonal antibody

Lieferant: United States Biological
USBIC5838-27-250EA 440 EUR
USBIC5838-27-250
Anti-CLDN3 Rabbit polyclonal antibody
Antikörper
Recommended dilution: Immunohistology: 1:50

Type: Primary
Antigen: CLDN3 (claudin 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR