Anti-CLDN2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: United States Biological
USBIC5838-16G-APEA 702 EUR
USBIC5838-16G-AP
Anti-CLDN2 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:250
Do not freeze!

Type: Primary
Antigen: CLDN2 (claudin 2)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR