Anti-CLDN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: United States Biological
USBIC5838-02F-APCEA 852 EUR
USBIC5838-02F-APC
Anti-CLDN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:50-1:100; Immunochemistry: 1:10-1:50
Do not freeze!

Type: Primary
Antigen: CLDN1 (claudin 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR